2,6-Dibromobenzenesulphonamide
Supplier: Apollo Scientific
Warning
OR55677-1G OR55677-250MG
APOSOR55677-1GEA 168 EUR
APOSOR55677-1G APOSOR55677-250MG
2,6-Dibromobenzenesulphonamide
2,6-Dibromobenzenesulphonamide
Formula: C₆H₅Br₂NO₂S
MW: 314.982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28783260
CAS Number: 1700401-95-1