2,6-Dimethoxy-3-nitropyridine 95+%
Supplier: Apollo Scientific
Warning
OR5849-1G OR5849-5G OR5849-10G
APOSOR5849-1GEA 42.2 EUR
APOSOR5849-1G APOSOR5849-5G APOSOR5849-10G
2,6-Dimethoxy-3-nitropyridine 95+%
2,6-Dimethoxy-3-nitropyridine
Formula: C₇H₈N₂O₄
MW: 184.149 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00832858
CAS Number: 18677-41-3