3-(Dimethylamino)piperidine 98+%
Supplier: Apollo Scientific
Warning
OR5899-1G OR5899-250MG OR5899-5G OR5899-100MG
APOSOR5899-1GEA 116 EUR
APOSOR5899-1G APOSOR5899-250MG APOSOR5899-5G APOSOR5899-100MG
3-(Dimethylamino)piperidine 98+%
3-(Dimethylamino)piperidine
Formula: C₇H₁₆N₂
Boiling Pt: 58 °C (6.75 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06796405
CAS Number: 50534-49-1