Isonicotinamidine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: Pyridine-4-carboxamidine hydrochloride, Isonicotinimidamide hydrochloride, 4-Carbamimidoylpyridine hydrochloride, 4-Amidinopyridine hydrochloride, Pyridine-4-carboximidamide hydrochloride, 4-Amidinopyridinium chloride
OR5939-1G OR5939-5G OR5939-25G
APOSOR5939-1GEA 21.9 EUR
APOSOR5939-1G APOSOR5939-5G APOSOR5939-25G
Isonicotinamidine hydrochloride
Isonicotinamidine hydrochloride
Formula: C₆H₇N₃·ClH
MW: 157.601 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00040385
CAS Number: 42518-06-9