(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one 97%
Supplier: Apollo Scientific
Warning
Synonyms: (1R,5S)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one, (1R,5S)-(+)-6,6-Dimethyl-2-oxobicyclo[3.1.1]heptane, (1R)-(+)-Nopinone
OR59892-1G OR59892-5G OR59892-10G OR59892-25G
APOSOR59892-1GEA 68.3 EUR
APOSOR59892-1G APOSOR59892-5G APOSOR59892-10G APOSOR59892-25G
(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one 97%
(1R,5S)-(+)-6,6-Dimethylbicyclo[3.1.1]heptan-2-one
Formula: C₉H₁₄O
MW: 138.207 g/mol
Boiling Pt: 209 °C
Density: 0.981 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08447116
CAS Number: 38651-65-9