4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Formylphenyl)-5-methyl-1,3,4-oxadiazole
OR59953-250MG
APOSOR59953-250MGEA 58 EUR
APOSOR59953-250MG
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde 95%
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde
Formula: C₁₀H₈N₂O₂
MW: 188.183 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD11109323
CAS Number: 179056-82-7