1-(tert-Butoxycarbonyl)-2-pyrrolidinone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(tert-Butoxycarbonyl)-2-oxopyrrolidine, tert-Butyl 2-oxopyrrolidine-1-carboxylate, Pyrrolidin-2-one, N-BOC protected
OR60029-1G OR60029-5G OR60029-25G OR60029-100G
APOSOR60029-1GEA 0 EUR
APOSOR60029-1G APOSOR60029-5G APOSOR60029-25G APOSOR60029-100G
1-(tert-Butoxycarbonyl)-2-pyrrolidinone
1-(tert-Butoxycarbonyl)-2-pyrrolidinone
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Boiling Pt: 100…105 °C (0.5 mm Hg)
Density: 1.086 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00674099
CAS Number: 85909-08-6