3-(Hexyloxy)propane-1,2-diol
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Hexyloxy)propylene glycol
OR60038-1G OR60038-5G
APOSOR60038-1GEA 109 EUR
APOSOR60038-1G APOSOR60038-5G
3-(Hexyloxy)propane-1,2-diol
3-(Hexyloxy)propane-1,2-diol
Formula: C₉H₂₀O₃
Boiling Pt: 109 °C (4 mm Hg)
Melting Pt: –5.5…0 °C
Density: 0.97 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00167017
CAS Number: 10305-38-1