Tetraethyl ethylene-1,1-diylbisphosphonate 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1-Bis(diethoxyphosphoryl)ethylene, 1,1-Bis(diethoxyphosphoryl)ethene, Tetraethyl vinylidene phosphonate, Tetraethyl ethene-1,1-diylbis(phosphonate)
OR60088-5G OR60088-1G
APOSOR60088-5GEA 357 EUR
APOSOR60088-5G APOSOR60088-1G
Tetraethyl ethylene-1,1-diylbisphosphonate 96%
Tetraethyl ethylene-1,1-diylbisphosphonate
Formula: C₁₀H₂₂O₆P₂
MW: 300.226 g/mol
Boiling Pt: 115…116 °C (0.05 mm Hg)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263518
CAS Number: 37465-31-9