3-(Aminomethyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazole trihydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: (6,7-Dihydro-5H-pyrrolo[2,1-c][1,2,4]triazol-3-yl)methylamine trihydrochloride, 6,7-Dihydropyrrolo[2,1-c][1,2,4]triazole-3-methylamine trihydrochloride
OR60095-1G OR60095-5G
APOSOR60095-1GEA 230 EUR
APOSOR60095-1G APOSOR60095-5G
3-(Aminomethyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazole trihydrochloride
3-(Aminomethyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazole trihydrochloride
Formula: C₆H₁₀N₄·3ClH
MW: 247.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699131
CAS Number: 1171236-13-7