1,3-Dimethyl-1H-benzimidazol-3-ium iodide
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dimethyl-1H-benzo[d]imidazol-3-ium iodide
OR61013-1G OR61013-250MG
APOSOR61013-1GEA 452 EUR
APOSOR61013-1G APOSOR61013-250MG
1,3-Dimethyl-1H-benzimidazol-3-ium iodide
1,3-Dimethyl-1H-benzimidazol-3-ium iodide
Formula: C₉H₁₁N₂·I
MW: 274.102 g/mol
Melting Pt: 137…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00491277
CAS Number: 7181-87-5