2,4,5-Trichlorobenzonitrile
Supplier: Apollo Scientific
Warning
OR61086-1G OR61086-250MG OR61086-5G
APOSOR61086-1GEA 24.8 EUR
APOSOR61086-1G APOSOR61086-250MG APOSOR61086-5G
2,4,5-Trichlorobenzonitrile
2,4,5-Trichlorobenzonitrile
Formula: C₇H₂Cl₃N
MW: 206.456 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787622
CAS Number: 6575-04-8