4-Formyl-3-hydroxybenzonitrile 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Cyano-2-formylphenol, 4-Cyano-2-hydroxybenzaldehyde

OR61317-250MG OR61317-1G
APOSOR61317-250MGEA 42.2 EUR
APOSOR61317-250MG APOSOR61317-1G
4-Formyl-3-hydroxybenzonitrile 95+%
4-Formyl-3-hydroxybenzonitrile
Formula: C₈H₅NO₂
MW: 147.131 g/mol
Melting Pt: 124…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08361770
CAS Number: 84102-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR