6-Bromoimidazo[1,2-a]pyridin-2-amine hydrochloride
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-6-bromoimidazo[1,2-a]pyridine hydrochloride
OR61410-1G OR61410-100MG OR61410-500MG
APOSOR61410-1GEA 492 EUR
APOSOR61410-1G APOSOR61410-100MG APOSOR61410-500MG
6-Bromoimidazo[1,2-a]pyridin-2-amine hydrochloride
6-Bromoimidazo[1,2-a]pyridin-2-amine hydrochloride
Formula: C₇H₆BrN₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD22205889
CAS Number: 1392102-12-3