4-Chloropyridine-3-boronic acid, pinacol ester hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine hydrochloride, 2-(4-Chloropyridin-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane hydrochloride
OR61415-1G OR61415-250MG OR61415-5G
APOSOR61415-1GEA 224 EUR
APOSOR61415-1G APOSOR61415-250MG APOSOR61415-5G
4-Chloropyridine-3-boronic acid, pinacol ester hydrochloride 97%
4-Chloropyridine-3-boronic acid, pinacol ester hydrochloride
Formula: C₁₁H₁₅BClNO₂·ClH
MW: 275.967 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18914522
CAS Number: 2102671-51-0