2-Bromo-4-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Danger
OR61542-1G OR61542-250MG
APOSOR61542-1GEA 255 EUR
APOSOR61542-1G APOSOR61542-250MG
2-Bromo-4-methoxybenzenesulphonyl chloride
2-Bromo-4-methoxybenzenesulphonyl chloride
Formula: C₇H₆BrClO₃S
MW: 285.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752806
CAS Number: 23095-16-1
UN: 3261
ADR: 8,III