7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine 97%
Supplier: Apollo Scientific
OR62035-1G OR62035-25G OR62035-5G
APOSOR62035-1GEA 21.9 EUR
APOSOR62035-1G APOSOR62035-25G APOSOR62035-5G
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine 97%
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine
Formula: C₆H₅IN₄
MW: 260.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21607515
CAS Number: 1770840-43-1