6-Ethynyl-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 6-Ethynyl-5-azaindazole, (1H-Pyrazolo[4,3-c]pyridin-6-yl)acetylene
OR62079-250MG OR62079-50MG OR62079-100MG
APOSOR62079-250MGEA 0 EUR
APOSOR62079-250MG APOSOR62079-50MG APOSOR62079-100MG
6-Ethynyl-1H-pyrazolo[4,3-c]pyridine
6-Ethynyl-1H-pyrazolo[4,3-c]pyridine
Formula: C₈H₅N₃
MW: 143.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22380516
CAS Number: 1374652-76-2