Methyl 3-methyl-1H-indole-5-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Methoxycarbonyl)-3-methyl-1H-indole
OR62081-1G OR62081-250MG
APOSOR62081-1GEA 516 EUR
APOSOR62081-1G APOSOR62081-250MG
Methyl 3-methyl-1H-indole-5-carboxylate
Methyl 3-methyl-1H-indole-5-carboxylate
Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11975645
CAS Number: 588688-33-9