17-Bromoheptadecanoic acid
Supplier: Apollo Scientific
Warning
OR6480-500MG OR6480-100MG
APOSOR6480-500MGEA 308 EUR
APOSOR6480-500MG APOSOR6480-100MG
17-Bromoheptadecanoic acid
17-Bromoheptadecanoic acid
Radiopharmaceutical precursor
Formula: C₁₇H₃₃BrO₂
MW: 349.347 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00040782
CAS Number: 13099-35-9
EINECS: 236-017-7