Methyl 17-bromoheptadecanoate
Supplier: Apollo Scientific
Warning
OR6489-100MG OR6489-250MG OR6489-200MG
APOSOR6489-100MGEA 433 EUR
APOSOR6489-100MG APOSOR6489-250MG APOSOR6489-200MG
Methyl 17-bromoheptadecanoate
Methyl 17-bromoheptadecanoate
Radiopharmaceutical precursor
Formula: C₁₈H₃₅BrO₂
MW: 363.373 g/mol
Boiling Pt: 212…214 °C (2.5 mm Hg)
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00040770
CAS Number: 72338-49-9