Methyl 16-bromohexadecanoate
Supplier: Apollo Scientific
Danger
Synonyms: Methyl16-bromohexadecanoate, Methyl 16-bromopalmitate
OR6490-1G OR6490-5G
APOSOR6490-1GEA 102 EUR
APOSOR6490-1G APOSOR6490-5G
Methyl 16-bromohexadecanoate
Methyl 16-bromohexadecanoate
Radiopharmaceutical precursor
Formula: C₁₇H₃₃BrO₂
MW: 349.347 g/mol
Melting Pt: 30…32 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040769
CAS Number: 26825-89-8