4'-Methyl-3'-nitroacetanilide
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-4-nitro-N-acetylbenzeneamine, N-(4-Methyl-3-nitrophenyl)acetamide
OR6637-1G OR6637-250MG OR6637-5G
APOSOR6637-1GEA 21.9 EUR
APOSOR6637-1G APOSOR6637-250MG APOSOR6637-5G
4'-Methyl-3'-nitroacetanilide
4'-Methyl-3'-nitroacetanilide
Formula: C₉H₁₀N₂O₃
MW: 194.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024277
CAS Number: 2719-14-4