Thieno[2,3-b]pyrazine-6-carboxylic acid 95%
Supplier: Apollo Scientific
Warning
OR6789-1G OR6789-250MG
APOSOR6789-1GEA 255 EUR
APOSOR6789-1G APOSOR6789-250MG
Thieno[2,3-b]pyrazine-6-carboxylic acid 95%
Thieno[2,3-b]pyrazine-6-carboxylic acid
Formula: C₇H₄N₂O₂S
MW: 180.184 g/mol
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD07371382
CAS Number: 59944-79-5
UN: 2811
ADR: 6.1,III