1-(4,4'-Dichlorobenzhydryl)piperazine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bis(4-chlorophenyl)methyl)piperazine, 1-[Bis(4-chlorophenyl)methyl]piperazine
OR6874-1G OR6874-5G OR6874-250MG
APOSOR6874-1GEA 43.5 EUR
APOSOR6874-1G APOSOR6874-5G APOSOR6874-250MG
1-(4,4'-Dichlorobenzhydryl)piperazine 98%
1-(4,4'-Dichlorobenzhydryl)piperazine
Formula: C₁₇H₁₈Cl₂N₂
MW: 321.244 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00191212
CAS Number: 27469-61-0
UN: 1759
ADR: 8 III