1-[3-(Diallylamino)prop-1-yl]piperazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: N,N-Diallyl-3-(piperazin-1-yl)propylamine, 1-{3-[Di(prop-2-en-1-yl)amino]prop-1-yl}piperazine
OR6918-25G OR6918-2G OR6918-5G
APOSOR6918-25GEA 477 EUR
APOSOR6918-25G APOSOR6918-2G APOSOR6918-5G
1-[3-(Diallylamino)prop-1-yl]piperazine 97%
1-[3-(Diallylamino)prop-1-yl]piperazine
Formula: C₁₃H₂₅N₃
MW: 223.358 g/mol
Boiling Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD01075180
CAS Number: 827614-52-8