Methyl 3-(4-chlorophenyl)-3-oxopropanoate 95+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl (4-chlorobenzoyl)acetate, Methyl 3-(4-chlorophenyl)-3-oxopropionate
OR6987-1G OR6987-5G OR6987-25G
APOSOR6987-1GEA 143 EUR
APOSOR6987-1G APOSOR6987-5G APOSOR6987-25G
Methyl 3-(4-chlorophenyl)-3-oxopropanoate 95+%
Methyl 3-(4-chlorophenyl)-3-oxopropanoate
Formula: C₁₀H₉ClO₃
MW: 212.63 g/mol
Boiling Pt: 132…138 °C (0.38 mm Hg)
Melting Pt: 42…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00216520
CAS Number: 22027-53-8