2'-Sulphamoyl-[1,1'-biphenyl]-4-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2-Sulphamoylphenyl)benzoic acid, 2'-(Aminosulphonyl)-[1,1'-biphenyl]-4-carboxylic acid
OR6989-1G OR6989-250MG
APOSOR6989-1GEA 615 EUR
APOSOR6989-1G APOSOR6989-250MG
2'-Sulphamoyl-[1,1'-biphenyl]-4-carboxylic acid
2'-Sulphamoyl-[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₃H₁₁NO₄S
MW: 277.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001764
CAS Number: 352615-90-8