3-Cyclopropylphenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (3-Hydroxyphenyl)cyclopropane
OR70007-1G OR70007-5G OR70007-250MG
APOSOR70007-1GEA 364 EUR
APOSOR70007-1G APOSOR70007-5G APOSOR70007-250MG
3-Cyclopropylphenol 97%
3-Cyclopropylphenol
Formula: C₉H₁₀O
MW: 134.175 g/mol
Boiling Pt: 80…82 °C (2 mm Hg)
Flash Pt: >70 °C
Storage Temperature: Ambient
MDL Number: MFCD06802406
CAS Number: 28857-88-7