4-Cyclopropylphenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (4-Hydroxyphenyl)cyclopropane
OR70008-250MG OR70008-1G
APOSOR70008-250MGEA 72.5 EUR
APOSOR70008-250MG APOSOR70008-1G
4-Cyclopropylphenol 98%
4-Cyclopropylphenol
Formula: C₉H₁₀O
MW: 134.175 g/mol
Boiling Pt: 83…85 °C (2 mm Hg)
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD06802405
CAS Number: 10292-61-2