3-Bromo-5-methylbenzene-1,2-diamine
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-methylphenylene-1,2-diamine, 3-Bromo-4,5-diaminotoluene
OR70018-1G OR70018-250MG OR70018-5G
APOSOR70018-1GEA 114 EUR
APOSOR70018-1G APOSOR70018-250MG APOSOR70018-5G
3-Bromo-5-methylbenzene-1,2-diamine
3-Bromo-5-methylbenzene-1,2-diamine
Formula: C₇H₉BrN₂
MW: 201.064 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18088524
CAS Number: 70733-25-4