Methyl 3-bromo-4,5-diaminobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-(methoxycarbonyl)phenylene-1,2-diamine, Methyl 3,4-diamino-5-bromobenzoate, 3-Bromo-5-(methoxycarbonyl)benzene-1,2-diamine
OR70021-1G OR70021-250MG
APOSOR70021-1GEA 29 EUR
APOSOR70021-1G APOSOR70021-250MG
Methyl 3-bromo-4,5-diaminobenzoate
Methyl 3-bromo-4,5-diaminobenzoate
Formula: C₈H₉BrN₂O₂
MW: 245.073 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18072579
CAS Number: 1245643-11-1