5-Chloronaphthalene-2-sulphonyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-2-(chlorosulphonyl)naphthalene
OR7150T-1G OR7150T-500MG
APOSOR7150T-1GEA 400 EUR
APOSOR7150T-1G APOSOR7150T-500MG
5-Chloronaphthalene-2-sulphonyl chloride
5-Chloronaphthalene-2-sulphonyl chloride
Formula: C₁₀H₆Cl₂O₂S
MW: 261.124 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Freezer
MDL Number: MFCD00269950
CAS Number: 89108-45-2
UN: 3261
ADR: 8,II