5,6-Dimethyl-4-hydroxy-2-thiopyrimidine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5,6-Dimethyl-2-sulphanylpyrimidin-4-ol, 5,6-Dimethyl-4-hydroxy-2-mercapto-1,3-diazine, 5,6-Dimethyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
OR7266-1G OR7266-5G OR7266-25G
APOSOR7266-1GEA 88.6 EUR
APOSOR7266-1G APOSOR7266-5G APOSOR7266-25G
5,6-Dimethyl-4-hydroxy-2-thiopyrimidine 95%
5,6-Dimethyl-4-hydroxy-2-thiopyrimidine
Formula: C₆H₈N₂OS
MW: 156.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269435
CAS Number: 28456-54-4