4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4'-Chloro-3-formylbiphenyl, 3-(4-Chlorophenyl)benzaldehyde, 4'-Chloro-[1,1'-biphenyl]-3-carbaldehyde
OR7356-1G
OR7356-5G
APOSOR7356-1GEA
79.9
EUR
APOSOR7356-1G
APOSOR7356-5G
4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde 95%
4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde
Formula:
C₁₃H₉ClO MW: 216.663 g/mol Melting Pt: 56…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD03424622 CAS Number: 139502-80-0 UN: 3077 ADR: 9+EHS III |