4-Phenylpyrimidine-2-thiol
Supplier: Apollo Scientific
Warning
OR7651-1G OR7651-5G
APOSOR7651-1GEA 139 EUR
APOSOR7651-1G APOSOR7651-5G
4-Phenylpyrimidine-2-thiol
4-Phenylpyrimidine-2-thiol
Formula: C₁₀H₈N₂S
MW: 188.249 g/mol
Melting Pt: 197…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00239517
CAS Number: 60414-59-7