Methyl 3-[(4,6-dimethylpyrimidin-2-yl)oxy]thiophene-2-carboxylate
Supplier: Apollo Scientific
Synonyms: 3-[(4,6-Dimethylpyrimidin-2-yl)oxy]-2-(methoxycarbonyl)thiophene
OR7902-1G OR7902-5G
APOSOR7902-1GEA 161 EUR
APOSOR7902-1G APOSOR7902-5G
Methyl 3-[(4,6-dimethylpyrimidin-2-yl)oxy]thiophene-2-carboxylate
Methyl 3-[(4,6-dimethylpyrimidin-2-yl)oxy]thiophene-2-carboxylate
Formula: C₁₂H₁₂N₂O₃S
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00203105
CAS Number: 389609-95-4