2-Amino-5-phenyl-3-furonitrile 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-phenylfuran-3-carbonitrile, 2-Amino-3-cyano-5-phenylfuran
OR8012-1G OR8012-250MG OR8012-5G
APOSOR8012-1GEA 66.9 EUR
APOSOR8012-1G APOSOR8012-250MG APOSOR8012-5G
2-Amino-5-phenyl-3-furonitrile 95%
2-Amino-5-phenyl-3-furonitrile
Formula: C₁₁H₈N₂O
MW: 184.194 g/mol
Melting Pt: 196…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00463472
CAS Number: 14742-32-6
UN: 3439
ADR: 6.1,III