3-(Aminomethyl)-2-bromothiophene hydrochloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: (2-Bromothien-3-yl)methylamine hydrochloride
OR8040-1G OR8040-250MG
APOSOR8040-1GEA 309 EUR
APOSOR8040-1G APOSOR8040-250MG
3-(Aminomethyl)-2-bromothiophene hydrochloride 97%
3-(Aminomethyl)-2-bromothiophene hydrochloride
Formula: C₅H₆BrNS·ClH
Boiling Pt: 70 °C (0.01 mm Hg)
Melting Pt: 199…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04971979
CAS Number: 886460-60-2
UN: 2922
ADR: 8+6.1,III