Hexamethoxyphosphazene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2,2,4,4,6,6-Hexamethoxy-1,3,5-triazatriphosphinine

OR8532-1G OR8532-250MG
APOSOR8532-1GEA 241 EUR
APOSOR8532-1G APOSOR8532-250MG
Hexamethoxyphosphazene 98%
Hexamethoxyphosphazene
Formula: C₆H₁₈N₃O₆P₃
MW: 321.145 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00436244
CAS Number: 957-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR