2-(4-Bromothien-2-yl)-1,3-dioxolane
Supplier: Apollo Scientific
Warning
OR8553-1G OR8553-5G
APOSOR8553-1GEA 21.9 EUR
APOSOR8553-1G APOSOR8553-5G
2-(4-Bromothien-2-yl)-1,3-dioxolane
2-(4-Bromothien-2-yl)-1,3-dioxolane
Formula: C₇H₇BrO₂S
MW: 235.098 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117938
CAS Number: 58267-85-9