6-Iodo-2-methyl-1,3-benzothiazole
Supplier: Apollo Scientific
Warning
Synonyms: 6-Iodo-2-methylbenzo[d]thiazole
OR8663-1G OR8663-250MG OR8663-5G
APOSOR8663-1GEA 105 EUR
APOSOR8663-1G APOSOR8663-250MG APOSOR8663-5G
6-Iodo-2-methyl-1,3-benzothiazole
6-Iodo-2-methyl-1,3-benzothiazole
Formula: C₈H₆INS
MW: 275.109 g/mol
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00464068
CAS Number: 68867-20-9
UN: 2811
ADR: 6.1 III