N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine 90%
Supplier: Apollo Scientific
Danger
Synonyms: 2-[2-(Dimethylamino)ethoxy]benzylamine, N-{2-[2-(Aminomethyl)phenoxy]ethyl}-N,N-dimethylamine
OR8685-1G OR8685-250MG
APOSOR8685-1GEA 309 EUR
APOSOR8685-1G APOSOR8685-250MG
N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine 90%
N,N-Dimethyl-2-[2-(aminomethyl)phenoxy]ethylamine
Formula: C₁₁H₁₈N₂O
MW: 194.273 g/mol
Boiling Pt: 89…91 °C (0.2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08690290
CAS Number: 91215-97-3
UN: 2735
ADR: 8,III