4'-Hydroxy-2'-methoxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Hydroxy-2-methoxyphenyl)ethanone, 4-Acetyl-3-methoxyphenol, 1-(4-Hydroxy-2-methoxyphenyl)ethan-1-one
OR8814-1G OR8814-250MG
APOSOR8814-1GEA 116 EUR
APOSOR8814-1G APOSOR8814-250MG
4'-Hydroxy-2'-methoxyacetophenone
4'-Hydroxy-2'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00038750
CAS Number: 493-33-4