7-(Benzyloxy)-DL-tryptophan
Supplier: Apollo Scientific
Warning
OR8850T-100MG OR8850T-10MG OR8850T-250MG
APOSOR8850T-100MGEA 277 EUR
APOSOR8850T-100MG APOSOR8850T-10MG APOSOR8850T-250MG
7-(Benzyloxy)-DL-tryptophan
7-(Benzyloxy)-DL-tryptophan
Formula: C₁₈H₁₈N₂O₃
Melting Pt: 234.5…236.0 °C
Storage Temperature: Freezer
MDL Number: MFCD00037969
CAS Number: 66866-40-8