Isopropyl toluene-4-sulphonate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Isopropyl 4-methylbenzenesulphonate, Prop-2-yl 4-methylbenzenesulphonate
OR8885-25G OR8885-5G OR8885-100G
APOSOR8885-25GEA 72.5 EUR
APOSOR8885-25G APOSOR8885-5G APOSOR8885-100G
Isopropyl toluene-4-sulphonate 97%
Isopropyl toluene-4-sulphonate
Formula: C₁₀H₁₄O₃S
MW: 214.281 g/mol
Melting Pt: 20 °C
Density: 1.142 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00968877
CAS Number: 2307-69-9
EINECS: 218-989-4