2-Bromoindane-1,3-dione 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-1H-indene-1,3(2H)-dione
OR8889-1G OR8889-5G OR8889-250MG
APOSOR8889-1GEA 256 EUR
APOSOR8889-1G APOSOR8889-5G APOSOR8889-250MG
2-Bromoindane-1,3-dione 96%
2-Bromoindane-1,3-dione
Formula: C₉H₅BrO₂
MW: 225.039 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00171530
CAS Number: 7319-63-3