1-Chloro-3-(methoxymethoxy)benzene 96%
Supplier: Apollo Scientific
OR900139-1G OR900139-5G
APOSOR900139-1GEA 219 EUR
APOSOR900139-1G APOSOR900139-5G
1-Chloro-3-(methoxymethoxy)benzene 96%
1-Chloro-3-(methoxymethoxy)benzene
Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18311888
CAS Number: 91105-99-6