N-Hydroxy-4-methylbenzenesulphonamide 97%
Supplier: Apollo Scientific
OR900172-1G OR900172-5G
APOSOR900172-1GEA 142 EUR
APOSOR900172-1G APOSOR900172-5G
N-Hydroxy-4-methylbenzenesulphonamide 97%
N-Hydroxy-4-methylbenzenesulphonamide
Formula: C₇H₉NO₃S
MW: 187.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11850835
CAS Number: 1593-60-8