2-chloro-1,3-dimethoxybenzene 95%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-1,3-dimethoxybenzene
OR900220-1G OR900220-5G
APOSOR900220-1GEA 21.9 EUR
APOSOR900220-1G APOSOR900220-5G
2-chloro-1,3-dimethoxybenzene 95%
2-chloro-1,3-dimethoxybenzene
Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835196
CAS Number: 7051-15-2